Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetraethylammonium bromide, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: Tetrylammonium bromide

TX0255-3 TX0255-3
EM-TX0255-3EA 542.43 USD InStock
EM-TX0255-3 CA-TX0255-3
Tetraethylammonium bromide, Sigma-Aldrich®
Tetraethylammonium bromide
Soluble in water.


Formula: C₈H₂₀BrN
MW: 210.16 g/mol
Melting Pt: > 285 °C
Density: 1.43 g/cm³ (20 °C)
MDL Number: MFCD00011825
CAS Number: 71-91-0
EINECS: 200-769-4
Merck Index: 12,09340
Order Now
Specification Test Results
Assay 99% min
Color White
Form Crystals
Infrared spectrum Conforms to standard

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty