Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iron(II) sulfate hydrate 86-89% (by manganometry) USP, BP, FCC, SAFC®

Supplier: MilliporeSigma
Warning
Synonyms: Ferrous sulfate hydrateFerrous sulphate hydrate

1.03967.9025
EM1.03967.9025EA 728 USD InStock
EM1.03967.9025
Iron(II) sulfate hydrate 86-89% (by manganometry) USP, BP, FCC, SAFC®
Iron (II) sulphate hydrate


Formula: FeSO₄·H₂O
MW: 169.93 g/mol
Melting Pt: ∼300 °C
Density: 2.97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149722
CAS Number: 13463-43-9
EINECS: 231-753-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty