Would you like to log in again?
KleenGuard® G10 Arctic Blue Nitrile Gloves, Kimberly-Clark
Supplier: Kimberly-Clark
KLEENGUARD®
90097 90095 90095 90097 90098 90099 90099 90096 90098 90096
89107-326CS 659.07 USD InStock
89107-326 CA89107-326 89107-328 CA89107-324 CA89107-328 89107-330 CA89107-332 89107-332 89107-324 CA89107-330
KleenGuard® G10 Arctic Blue Nitrile Gloves, Kimberly-Clark
Gloves
Powder-free, latex-free gloves are more comfortable and economical than standard nitrile gloves, yet retain the protective properties required for excellent performance across a wide range of industrial applications.
  • Ambidextrous
  • Beaded cuff
  • Textured fingertips
Ideal replacement for latex gloves.
Certifications: Food handling certified in North America and Europe. AQL 1.5.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty