Would you like to log in again?
BD IV Start Pak™ Kits, BD Medical
Supplier: BD Medical
BD IV Start Pak™
386159 386150 386142 386120
63019-857CS 739.87 USD InStock
63019-857 63019-869 BD386150 63019-855
BD IV Start Pak™ Kits, BD Medical
Catheters
These sterile, single-use kits provide all the components needed for an efficient and effective peripheral catheter insertion
All kits include a set of IV accessories: one alcohol wipe, one tourniquet, two 5x5cm (2x2”) gauze sponges, one roll of Transpore™ tape, and one ID label. The Basic Kit also includes one adhesive bandage. The Basic Tegaderm® kit includes one frame-style Tegaderm® IV dressing. The Drape Kit includes one frame-style Tegaderm® dressing and one impermeable drape. The Glove Kits include a Tegaderm® dressing and one set of medium exam gloves.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty