Would you like to log in again?
VWR® Disposable Gloves, PE
Supplier: Avantor
VWR®
2GM2VW 2GELBV 2GM1VW 2GM3VW
32915-272CS 663.17 USD InStock
32915-272 32915-246 32915-224 32915-268
VWR® Disposable Gloves, PE
Gloves
Use these gloves for excellent hand and product protection in virtually any type of laboratory, food handling, industrial safety, veterinary, electronics, painting, printing, or photography environment.
  • Ultra soft and pliable
  • Tough, waterproof construction
  • Resistant to most chemicals and detergents
  • Extra-long cuffs provide added protection
  • Convenient pull-out box dispenser
  • Ambidextrous, embossed finish
Soft, virgin polyethylene gloves are so pliable you can handle even small laboratory apparatus with ease. Resistant to most chemicals or detergents. Ambidextrous. Embossed finish.
Certifications: Gloves are made using only listed materials and levels outlined in CFR 21 170-199.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty