Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PIPES monosodium salt, clear, colorless, crystalline powder

Supplier: VWR
Synonyms: PIPES-Na2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid monosodium saltPiperazine-1,4-bis(2-ethanesulphonic acid) monosodium saltSodium hydrogen piperazine-1,4-diethanesulfonatePiperazine-N,N'-bis-2-ethanesulfonic acid monosodium saltMonosodium piperazine-1,4-diethanesulphonatePiperazine-1,4-bis(2-ethanesulfonic acid) monosodium saltSodium 2-(4-(2-sulfoethyl)piperazin-1-yl)ethanesulfonate2,2'-(Piperazine-1,4-diyl)bis(ethanesulfonic) acid monosodium saltPiperazine-N,N'-bis-2-ethanesulphonic acid monosodium saltSodium hydrogen piperazine-1,4-diethanesulphonateMonosodium piperazine-1,4-diethanesulfonate

VWRV0169-5KG VWRV0169-10KG VWRV0169-50KG VWRV0169-1KG
77437-882EA 70925.8 USD InStock
77437-882 80760-038 80760-036 80760-034
PIPES monosodium salt, clear, colorless, crystalline powder
PIPES monosodium salt


Formula: C₈H₁₇N₂NaO₆S₂
MW: 324.35 g/mol
Storage Temperature: Room Temperature
MDL Number: MFCD00065472
CAS Number: 10010-67-0
Order Now
Specification Test Results
Assay ≥99% (titration)
pH 4.5 - 4.7
pH range 6.1 - 7.5
pKa (25 °C) 6.8
Solubility 0.1 M NaOH: 250 mg/ml

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty