Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Mercaptoethanesulfonic acid sodium salt (mesna) ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: Sodium 2-mercaptoethanesulfonateSodium 2-mercaptoethanesulphonate

M0913-25G M0913-10G
TCM0913-010GEA 138.52 USD InStock
TCM0913-010G TCM0913-25G
2-Mercaptoethanesulfonic acid sodium salt (mesna) ≥95.0% (by titrimetric analysis)
2-Mercaptoethanesulphonic acid sodium salt (Mesna)
CAS Number: 19767-45-4
MDL Number: MFCD00007535
Molecular Formula: C2H6O3S2
Molecular Weight: 164.17
Purity/Analysis Method: 95.0% (T)
Form: Crystal

Product Categories: Bioscience, Antitumor Ingredients
Other Product Names / Synonyms: 2-Mercaptoethanesulfonic Acid Sodium Salt, Mesna


Formula: C₂H₅NaO₃S₂
MW: 164.18 g/mol
MDL Number: MFCD00007535
CAS Number: 19767-45-4
EINECS: 243-285-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/44/53/28974453.jpg
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty