Would you like to log in again?
Cryo-Gloves Industrial® Low Temperature Gloves
Supplier: Tempshield
Cryo-Gloves®
CIGMWP CIMMWP CIWMWP CIGXWP
89511-894CS 2280.56 USD InStock
89511-894 89511-910 89511-900 89511-906
Cryo-Gloves Industrial® Low Temperature Gloves
Gloves
Cryo-Industrial® Gloves protect your hands and arms when working in hazardous, ultra-cold environments.
  • Maximum thermal protection
  • Waterproof
  • Lightweight and comfortable
  • Excellent dexterity
  • Breathable and stay-dry lining
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty