Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

meta-Phosphoric acid stabilized, GR ACS, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Metaphosphoric acid

PX1004-1 PX1004-5
EM-PX1004-5EA 7390 USD InStock
EM-PX1004-5 EM-PX1004-1
meta-Phosphoric acid stabilized, GR ACS, Supelco®
meta-Phosphoric acid
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs


Formula: (HPO₃)n
MW: 79.98 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 21 °C
Density: 2.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011351
CAS Number: 37267-86-0
EINECS: 253-433-4
UN: 3260
ADR: 8,III
Order Now
Specification Test Results
Assay (HPO3) 33.5-36.5%
Arsenic (As) 1ppm max
Chloride (Cl) 0.001% max
Color White or clear
Form Crystals/lumps
Heavy metals (as Pb) 0.005% max
Iron (Fe) 0.005% max
Nitrate (NO3) 0.001% max
Stabilizer (NaPO3) 57.0-63.0%
Substances reducing permanganate (as H3PO3) 0.02% max
Sulfate (SO4) 0.005% max

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty