Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid solution 10% (w/w) FCC

Supplier: Columbus Chemical Industries, Inc
Warning

986900-22501 986900-21001
95000-002EA 2010.52 USD InStock
95000-002 95000-000
Sulfuric acid solution 10% (w/w) FCC
Sulfuric acid


Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 101 °C (1013 hPa)
Density: 1.066 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II
Merck Index: 13,09064
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty