Would you like to log in again?
Apiezon® Sealing Compound Q
Supplier: M & I Materials
Apiezon®
Q-01000
10769-162EA 498.06 USD InStock
10769-162
Apiezon® Sealing Compound Q
Lubricants
Putty-like sealing compound used to fill gaps and seal joints on the rotary pump side of a vacuum system; forms seals around flat, unground joints
The sealing compound softens between 40–50°C (104–122°F). Maximum temperature for use is 30°C (86°F). Vapor pressure is 1x10-4 Torr at 20°C (68°F).
Constructs temporary vacuum enclosures (such as sealing glass cylinders to metal plates). Used for vacuum work where low vapor pressures are not required. Readily molded. Easily applied and removed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty