Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride 99-100.5%, granular, AR®, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: HaliteSea saltSaline

Management of Change (MOC) category = N

MACR7544-06 MACR7544-06
MK754406CS 1127.57 USD InStock
CAMK754406 MK754406
Sodium chloride 99-100.5%, granular, AR®, Macron Fine Chemicals™
Sodium chloride
Granular.


Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (NaCl) (dried basis) 99.5 - 100.5 %
Appearance (white, crystalline granules) Passes Test
Solubility (10 in 50) Passes Test
Identification A Passes Test
Identification B Passes Test
Loss on Drying at 105°C ≤0.5 %
Acidity or Alkalinity Passes Test
Limit of Bromide ≤0.010 %
Calcium and Magnesium (as Ca) ≤0.005 %
Magnesium and Alkaline Earth Metals (as Ca) ≤0.01 %
Phosphate (PO₄) ≤0.0025 %
Barium (Ba) Passes Test
Arsenic (As) ≤0.0001 %
Heavy Metals (as Pb) ≤0.0005 %
Iron (Fe) ≤0.0002 %
Sulfate (SO₄) ≤200 ppm
Iodide (I) Passes Test
Copper (Cu) ≤0.0002 %
Sodium Ferrocyanide Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty