Would you like to log in again?
SP Bel-Art Bellows Type Glove Box Gloves, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
F50025-0331 H50025-0310 500250309 F50025-0332 F50025-0333
74950-356EA 98.04 USD InStock
74950-356 47735-514 47735-516 74950-360 74950-358
SP Bel-Art Bellows Type Glove Box Gloves, Bel-Art Products, a part of SP
Gloves
Made from natural latex with a removable hand, the sleeves fit 20 cm (8") circular ports and 18×25 cm (7×10") oval ports.
  • Natural latex construction
  • Can house any sized gloves
  • Leakproof
  • Bellows allow cooling air circulation
The length of the extended sleeve and glove is approximately 76 cm (30").
When the glove and sleeve are separated, the sleeve remains attached to the box. Glove sizes are interchangeable, and almost any standard laboratory, surgical or disposable glove can be used.
Forms a leak-proof unit with O-ring construction integral with the box. The bellows allows cooling air circulation. Use with 47748-192 Clamping rings to hold gloves in place.
Ordering information: Gloves are sold by the pair.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty