Would you like to log in again?
Kevlar® Cut-Resistant String Knit Gloves, Wells Lamont
Supplier: Wells Lamont
Kevlar®
1800M 1800L 1800XL 1800S
80091-344EA 15.97 USD InStock
80091-344 80091-342 80091-340 80091-338
Kevlar® Cut-Resistant String Knit Gloves, Wells Lamont
Gloves
These 100% Kevlar string-type gloves offer high tensile strength, low density, and resistance to corrosion.
  • Lightweight
  • Ambidextrous
  • Reinforced thumb stitch
  • Continuous cuff
  • Corrosion resistant
Gloves provide cut resistance without sacrificing comfort and dexterity and have a reinforced thumb stitch and a continuous cuff.
The lightweight gloves are ambidextrous.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty