Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrabutylammonium hydroxide solution 25% in methanol, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Management of Change (MOC) category = N

BAKRV362-05 BAKRV362-05
JTV362-5EA 444.2 USD InStock
JTV362-5 CAJTV362-5
Tetrabutylammonium hydroxide solution 25% in methanol, BAKER ANALYZED®, J.T.Baker®
Tetrabutylammonium hydroxide
Titrant. Lot analysis on label.


Formula: C₁₆H₃₇NO
MW: 259.48 g/mol
MDL Number: MFCD00009425
CAS Number: 2052-49-5
UN: 3263
ADR: 8,II
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Molarity 0.98 - 1.08
1.0 M Tris Base
1.0 M Acetate
0.025 M EDTA
Filtered through a 0.2 micron filter.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty