Synonyms:
Di-p-tolyl sulphoxide
, Di-p-tolyl sulfoxide
T1075-25G
TCT1075-025GEA
286
USD
InStock
TCT1075-025G
p-Tolyl sulfoxide ≥98.0% (by GC)
p-Tolyl sulfoxide
CAS Number: 1774-35-2
MDL Number: MFCD00008546
Molecular Formula: C14H14OS
Molecular Weight: 230.33
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 94
Other Product Names / Synonyms: Di-p-tolyl Sulfoxide