Would you like to log in again?
Linearity LQ Ammonia/Ethanol for Abbott Systems
Supplier: AUDIT Microcontrols
K899M-5
77392-052EA 480.77 USD InStock
77392-052
Linearity LQ Ammonia/Ethanol for Abbott Systems
Control reagents
Linearity LQ ammonia/ethanol for abbott systems is intended to simulate human patient serum samples for the purpose of determining linearity, calibration verification, and verification of reportable range for ammonia and ethanol.
  • Linearity liquid
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty