Would you like to log in again?
Secovac 142 Clear Static Dissipative Bags, 3 mil, SECO
Supplier: SECO
Secovac
142Z-1012 142Z-35 142Z-46 142-68 142Z-68 142Z-810 142-1215 142-1518 142-46 142Z-1215 142-35 142-1012 142-810
14218-040PK 178.96 USD InStock
14218-040 14218-050 14218-042 14218-030 14218-052 14218-048 14218-036 14218-028 14218-038 14218-044 14218-032 14218-046 14218-034
Secovac 142 Clear Static Dissipative Bags, 3 mil, SECO
Bags Sample Bags
Amine-free, mono-extruded formulation that is polycarbonate compatible, humidity independent, non-octanoic.
  • Non-corrosive and non-contaminating
Polyethylene bags are hot stamped with blue ESD warning symbol.
Certifications: Tested to meet or exceed the electrical and physical requirements of MIL-B-81705C Type II and EIA 541.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty