Would you like to log in again?
RoadMaster Leather Palm Driving Gloves, Magid
Supplier: Magid
B540L B540M
69100-033PK 215.84 USD InStock
69100-033 69100-031
RoadMaster Leather Palm Driving Gloves, Magid
Gloves
The RoadMaster is a tan, cow grain, unlined all-leather drivers’ glove with a gunn cut pattern and a straight thumb.
  • Premium grain leather, unlined drivers glove
  • Ball and tape closure holds glove securely
  • Straight thumb, gunn cut pattern
  • Ideal for a variety of uses
  • Dependable RoadMaster brand
It cow grain driving gloves are an ideal glove for drivers, utility workers, warehouse employees, light welding and assembly workers.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty