Would you like to log in again?
Clear Lens Replacement, 611 Polycarbonate (AF121 Treated)
Supplier: Univet
042.120M
77393-704CS 10350 USD InStock
77393-704
Clear Lens Replacement, 611 Polycarbonate (AF121 Treated)
Goggles
611 Polycarbonate AF121 spare lens for use with the 611 series frame.
  • Clear replacement lens
  • Lens treatment: anti-fog and anti-scratch
  • Optical class (1)
  • Lens mark (EU): 2C-1.2 U 1 BT CE
Replacement polycarbonate lens for 611 series goggles is reusable for up to 40 autoclave sterilization cycles with steam at 121 °C, 1 atm for 30 minutes. The AF121 treatment applied to the lens prevents scratches and fogging at the same time. The lens will withstand up to 10 steam sterilization cycles. It is also reusable for up to 10 gamma irradiation cycles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty