Would you like to log in again?
Heat-Resistant Terrycloth Gloves, Wells Lamont
Supplier: Wells Lamont
1966
80091-274EA 9.5 USD InStock
80091-274
Heat-Resistant Terrycloth Gloves, Wells Lamont
Gloves
Heavyweight, seamless gloves feature superior cut and abrasion protection.
  • Seamless
  • White
An insulating layer of air forms beneath each loop of fabric, allowing the gloves to dissipate heat quickly.
The ambidextrous, knit-wrist gloves protect against up to 176 °C (350 °F).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty