Would you like to log in again?
Secovac 142 Bags, SECO
Supplier: SECO
Secovac
142-1212 142Z-68 142-68 142Z-1012 142-46 142Z-810 142Z-1012
58605-924PK 33.1 USD InStock
58605-924 58605-902 58605-964 58605-962 58612-000 58605-928 58605-926
Secovac 142 Bags, SECO
Bags Sample Bags
Static-dissipative bags ensure high ESD protection and are reusable without retesting.
  • Designed for use in all ranges of temperature and humidity
  • Offers low ionic contamination and permanence of static-dissipative properties
  • Made with polyolefin copolymer resins
  • Available in zip-close or open-top styles
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty