Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Phenyl-3-(trimethylsilyl)-2-propyn-1-ol ≥95%

Supplier: AOBChem USA

Warning
Synonyms: α-[2-(Trimethylsilyl)ethynyl]benzenemethanol

277527-25G 277527-5G
77491-046EA 95 USD InStock
77491-046 77501-096
1-Phenyl-3-(trimethylsilyl)-2-propyn-1-ol ≥95%
1-Phenyl-3-(trimethylsilyl)-2-propyn-1-ol
Caution: For research use only


Formula: C₁₂H₁₆OSi
MW: 204.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05864336
CAS Number: 89530-34-7
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty