Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Decafluorobenzophenone ≥95%

Supplier: AOBChem USA

Warning
Synonyms: PerfluorobenzophenoneDecafluorobenzophenone

318886-5G
77492-132EA 114 USD InStock
77492-132
Decafluorobenzophenone ≥95%
Decafluorobenzophenone
Caution: For research use only


Formula: C₁₃F₁₀O
MW: 362.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000295
CAS Number: 853-39-4
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty