Would you like to log in again?
NIGHTSEA® Light Head and Filter Sets
Supplier: Electron Microscopy Sciences
NIGHTSEA®
SFA-LFS-GR SFA-LFS-UV SFA-LFS-VI SFA-LFS-CY SFA-LFS-RB-GO
77463-322EA 1128.47 USD InStock
77463-322 77463-320 102773-524 102773-522 77463-318
NIGHTSEA® Light Head and Filter Sets
Lights Fluorescent Lights, Lamps and Bulbs
Sets consist of matched NIGHTSEA® light head, barrier filter, and viewing shield in a padded storage box.
  • Light head
  • Barrier filter
  • Viewing shield
  • Padded storage box
Compatible with the NIGHTSEA® SFA stereo microscope fluorescence adapter system.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty