Would you like to log in again?
PYREX® Single Metric Scale Graduated Cylinders, To Contain, Corning
Supplier: Corning
PYREX®
3012-250 3012-100 3012-50
24763-240CS 76.73 USD InStock
24763-240 24763-266 24763-244
PYREX® Single Metric Scale Graduated Cylinders, To Contain, Corning
Cylinders Measuring Cylinders
Borosilicate glass, without stoppers.
  • Calibrated 'to contain' (TC)
  • Equipped with a 24/40 ST joint
  • Single metric scale
  • Graduations are in durable white enamel
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty