Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Phenyl-1-pentyn-3-ol ≥96%

Supplier: AOBChem USA

Warning
Synonyms: 1-Phenyl-1-pentyn-3-ol

258767-25G 258767-5G
77500-728EA 394 USD InStock
77500-728 77490-594
1-Phenyl-1-pentyn-3-ol ≥96%
1-Phenyl-1-pentyn-3-ol
Caution: For research use only


Formula: C₁₁H₁₂O
MW: 160.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041589
CAS Number: 27975-78-6
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty