Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fura-2 LR/AM ≥95% (by HPLC), Calbiochem®

Supplier: MilliporeSigma
Synonyms: Fura-2 Leakage Resistant (AM)

344911-1SET 344911-1SET
80051-796EA 398.55 USD InStock
CA80051-796 80051-796
Fura-2 LR/AM ≥95% (by HPLC), Calbiochem®
Fura-2 LR/AM
Acetoxymethyl (AM) derivative of FURA-PE3 (80051-794) with spectral properties similar to the parent compound

Can be loaded into cells in a manner similar to FURA 2/AM (80051-788). Hydrolysis by non-specific esterases traps the indicator in the cytosol. Excitation maximum: 364nm (low Ca2+) and 335nm (high Ca2+); emission maximum: 508nm (low Ca2+) and 495nm (high Ca2+).


Formula: C₅₅H₆₃N₅O₂₉
CAS Number: 172890-84-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty