Would you like to log in again?
VWR® Data Logger
Supplier: Avantor
VWR®
4325
21800-026EA 935.81 USD InStock
21800-026
VWR® Data Logger
Data Loggers
Data can be read as-is, or imported to spreadsheets, databases, and statistical programs.
Complete DAS–4™ captures and stores up to 8000 bytes (over 1000 readings). Readings may be taken at intervals from 1 second to 99 hours. Stored readings may be downloaded to any PC and viewed. For use with Traceable™ meters featuring an RS232 output, computer output.
Ordering information: Supplied with 91.4 cm (36") serial cable with D9F computer plug, 8.9 cm (3¹/₂") Windows® diskette, and four AA alkaline batteries.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty