Would you like to log in again?
VWR® Boston Round Bottles, Amber, Narrow Mouth
Supplier: Avantor
VWR®
VW5120120V-21 VW5123233V-26 VW5120120V-26 VW5120824V-21 VW5123233V-22 VW5121628V-26 VW5123233C-24 VW5120120C-25 VW5120422V-26 VW5123233B VW5120120V-25
89042-910CS 123.68 USD InStock
89042-910 89042-914 89042-916 89042-906 89042-792 89042-880 89042-882 89042-894 89042-786 89042-940 89042-820
VWR® Boston Round Bottles, Amber, Narrow Mouth
Bottles Glass Bottles
Bottles provide an ideal combination of quality and economy.
  • Soda-lime glass fabrication
  • Amber tint protects against UV light
Constructed of Type III soda-lime glass, the amber tinting provides protection for light-sensitive products and samples. Bottles are supplied with black phenolic caps (with the exception of all items designated 'Bottles Only').
Ordering information: Convenience packs consist of corrugated cases with dividers separating glassware in smaller quantities (12 to 48 per pack). Bulk packs consist of shrink-wrapped glassware bundles (12 to 432 per wrap) packaged in corrugated cases. Bottle caps are included in separate bags within the case. Bottles sold without caps are packaged in bulk packs. Pallet packs consist of shrink-wrapped glassware bundles stacked onto shrink-wrapped pallets. This packaging is designed for large glassware quantities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty