Would you like to log in again?
Diamond® RealSeal™ Bottles, Wide Mouth, Square, HDPE, Amber
Supplier: Globe Scientific
Diamond® RealSeal™
7180500AM 7180250AM 7180175AM 7181000AM 7180060AM
77448-014PK 110.02 USD InStock
77448-014 77448-012 77448-010 77448-008 77448-016
Diamond® RealSeal™ Bottles, Wide Mouth, Square, HDPE, Amber
Bottles Plastic Bottles
Diamond RealSeal™ Square plastic laboratory bottles from Globe Scientific meet the requirements of the most demanding applications.
  • Reduce UV Light transmission to protect light sensitive contents during storage and transport
  • Comply with US Pharmacopeia requirements for maximum light transmission
  • Safe for freezer storage down to –100 °C
  • Resistant to most corrosives
  • HDPE with PP Closure
These new additions to the Diamond RealSeal offering expand the range with a selection of bottles that store and pack more efficently than standard round bottles. They are available in several material options.
These new bottles are precision molded from virgin resins for consistent wall thickness and smooth interior and exterior surfaces. The bottles include leak-proof polypropylene caps that are engineered with an integrated seal ring for leak-proof closure.
These square bottles store and pack efficiently to maximize space utilization. The wide mouths are perfect for sample collection and facilitate the rapid filling and emptying of solids, powders, and viscous liquids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty