Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
PTFE Vial Funnels, Chemglass
Supplier: Chemglass
CG-1761-05 CG-1761-02 CG-1761-04 CG-1761-07 CG-1761-06 CG-1761-01 CG-1761-08 CG-1761-03 CG-1761-20
89049-894EA 38.5 USD InStock
89049-894 89049-884 89049-892 89049-898 89049-888 89049-896 89049-886 89049-900 89049-890
PTFE Vial Funnels, Chemglass
Funnels Filtration Funnels Filter Funnels
Fluoropolymer resin funnels transfer contents safely, and screw securely onto any standard laboratory vial with a GPI thread.
Ordering information: Funnel kit includes polypropylene box with one of each of the eight funnel sizes listed below.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty