Would you like to log in again?
Embedding Cassettes, Strip Holed, Two-Piece
Supplier: Four E's USA
K2106040 K2106041
76809-036CS 712.89 USD InStock
76809-036 76809-038
Embedding Cassettes, Strip Holed, Two-Piece
Embedding Cassettes Tissue Processing Cassettes
POM embedding cassettes ensure precise tissue processing with minimized deformation.
  • POM material minimizes wax block deformation
  • Even and uniform printing surfaces
  • Front-hinged design allows easy cover removal
  • 45° angled writing surface for convenient labeling
  • For use with laser printers
Crafted from POM material, these embedding cassettes offer exceptional durability and minimize wax block deformation during tissue processing. Printing and labeling surfaces are perfectly flat, enabling high-quality labeling with text and data matrix codes. These cassettes offer a front-hinged design (also known as reverse hinged). The detachable cover can be easily removed and re-attached as many times as desired.
Packaging: Sleeved packaging: 90 per sleeve, 20 sleeves per case. Bulk packaging: 500 per bag, 6 bags per case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty