Would you like to log in again?
Diamond® Essentials™ Bulk Wide Mouth HDPE Bottles
Supplier: Globe Scientific
Diamond® Essentials™
7510125BKS 7510030BKS 7510060BKS 7510250BKS 7510500BKS
77506-732CS 667.2 USD InStock
77506-732 77506-734 77506-740 77506-736 77506-738
Diamond® Essentials™ Bulk Wide Mouth HDPE Bottles
Bottles Plastic Bottles
Diamond® Essentials™ bulk plastic bottles meet the market need for efficient, effective, and economical packaging options aimed at manual and automated filling operations.
  • Manufactured in ISO 13485 and ISO 9001 certified facility
  • Medical grade resins to meet demanding requirements
  • Molded to precise specifications for consistent wall thickness and smooth surfaces
  • Leakproof, linerless caps for secure closure
  • Industry accepted cap and closure sizes for seamless integration into filling operations
Diamond® Essentials™ bulk plastic laboratory bottles are precision molded from virgin high density polyethylene (HDPE) resins for consistent wall thickness and smooth surfaces inside and out. These rugged HDPE bottles are resistant to most corrosives and are suitable for freezer storage down to −100 °C. Polypropylene caps are engineered with an integrated seal ring and semi-buttressed threads for leakproof closure. These leakproof bottles are designed and tested to withstand the highest pressure thresholds in the industry.
Packaged in bulk with caps and bottles bagged separately, no de-capping step is needed before filling Diamond® Essentials™ Bulk Bottles. Available in wide and narrow mouth options with cap and closure sizes that match industry norms, the bottles integrate easily into filling operations.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty