Would you like to log in again?
PYREX® Burets, Serialized and Certified, Class A, Colored Scale, Straight Bore, Fluoropolymer Resin Stopcock Plug, Corning
Supplier: Corning
PYREX®
2105-100 2105-50 2105-25 2105-10
17473-078CS 381.57 USD InStock
17473-078 17473-084 17473-075 77776-974
PYREX® Burets, Serialized and Certified, Class A, Colored Scale, Straight Bore, Fluoropolymer Resin Stopcock Plug, Corning
Burets
Borosilicate glass, class A.
  • Individually serialised
  • Colored markings with fine sharp lines
  • Large easy to read numbers
  • Smooth micro-finish stopcock assures leak-resistant performance
Buret tips are carefully drawn from precision bore tubing to ensure proper drainage rates.
Designed as per ASTM E287 and calibrated in accordance with ASTM E542 and D-511 requirements. Traceable to NIST standards.
Certifications: PYREX® Serialized/Certified Volumetric Glassware is now tested and calibrated in an ISO/IEC 17025 certified laboratory.
Delivery information: Burets are supplied with 2 mm bore PTFE stopcock plug, a dust cover and certificate of identification and capacity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty