Would you like to log in again?
ThermaSeal™ 60 Wipers, Texwipe®
Supplier: Texwipe
ThermaSeal™
TX2064 TX2069
TWTX2064CS 804.69 USD InStock
TWTX2064 TWTX2069
ThermaSeal™ 60 Wipers, Texwipe®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Hot-cut polyester wipers for all-purpose cleaning in ISO Class 3 (FED-STD-209E Class 1/M1.5) critical cleanroom environments.
  • Good absorbency
  • Wiping efficiency
  • Abrasion resistance
Wipers are made of 100% continuous-filament, double-knit fabric with thermally sealed edges for fiber control.
Ordering information: Laundered and packaged in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom environment. Packed in SolventSafe Bag-Within-A-Bag® cleanroom packaging.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty