Would you like to log in again?
SP Wilmad-LabGlass Gooch Crucibles, High Form, SP Industries
Supplier: SP Industries
LG-5470-108 LG-5470-102 LG-5470-100
80089-456EA 75.43 USD InStock
80089-456 80089-448 80089-450
SP Wilmad-LabGlass Gooch Crucibles, High Form, SP Industries
Crucibles
With fused-in fritted glass disc, these high form borosilicate glass Gooch crucibles are designed for analytical work involving precipitation, drying, and weighing.
They are particularly suitable when precipitates will not be dried at temperatures above 110°C, and may be used at higher temperatures. Fine (F), Medium (M), and Coarse (C) porosities available.
Caution: Note: Avoid sudden temperature changes when heating or cooling.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty