Would you like to log in again?
ESD-Safe Sheet Protectors, SECO
Supplier: SECO
SP142-1711 SP142-811 SP142-811
14223-014CS 983.57 USD InStock
14223-014 58605-522 58605-948
ESD-Safe Sheet Protectors, SECO
Sheet Protectors
ESD-Safe Sheet Protectors are manufactured with coextruded, multilayer Secovac 142 polypropylene film.
  • Sealed radius corners
  • Edges provide strength, durability, and protection
  • Easy-to-read
  • Non-glare finish
  • Die cut for three-ring binders
  • Resistant to photocopy
These sheet protectors are static-dissipative. They feature noncontaminating properties, are polycarbonate compatible.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty