Would you like to log in again?
TOC X5 Orbital Shakers for TNTplus Tests, Hach
Supplier: Hach
LQV1489900002 LQV148.99.00002
75921-232EA 3072.2 USD InStock
75921-232 104087-498
TOC X5 Orbital Shakers for TNTplus Tests, Hach
Shakers and Mixers Orbital Shakers
The TOC X5 Shaker is required for the TNT810 and TNT811 TOC TNTplus tests. It purges the Total Inorganic Carbon in the vial, which means you do not have to use or clean any lab glassware.
  • Mains Connection 3: 30 VA
  • Manual Languages: English, French, Spanish, Portugese, Chinese
  • Parameter: TOC (Total Organic Carbon)
  • Warranty: 12 months
The TOC X5 Shaker allows to prepare eight samples in parallel in only five minutes, shortening the time spent on your analysis.
Delivery information: Includes: TOC-X5 Shaker, plug-in power supply unit, non-slip mat.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty