Would you like to log in again?
Spordex® VH202 Biological Indicator Disc
Supplier: Steris
NA333
77463-572PK 950 USD InStock
77463-572
Spordex® VH202 Biological Indicator Disc
Sterilization Indicators
Spordex® VH202 stainless steel biological indicator is used in cycle development and routine monitoring of applications with STERIS VHP biodecontamination units.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty