Would you like to log in again?
PRISM® 410™ Black Toughened Instant Adhesive, Loctite®, Henkel
Supplier: Henkel
PRISM® 410™
41045
82002-794CS 358.2 USD InStock
82002-794
PRISM® 410™ Black Toughened Instant Adhesive, Loctite®, Henkel
Adhesives
This black, toughened ethyl adhesive bonds rubbers, metals, and plastics for use in difficult environments.
  • Tough adhesive
  • For use in difficult environments
  • Fills gaps up to 0.020cm (0.008")
Adhesive is peel- and shock-resistant.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty