Would you like to log in again?
EPA method 502.2 Standards Kit, Restek
Supplier: Restek
30041 30040
RK30041EA 39.77 USD InStock
RK30041 RK30040
EPA method 502.2 Standards Kit, Restek
Standards Environmental Analysis Standards
Standards are packaged in purge-and-trap-grade methanol at a concentration of 2000 µg/mL in a 1 mL ampoule.
  • Various internal standards available
Individual VOA surrogate and internal standards are suitable for EPA methods.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty