Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Acid Matrix Spike Standard, Restek
Supplier: Restek
31061 31071 31061-15
75834-606PK 605.44 USD InStock
75834-606 RK31071 RK31061
Acid Matrix Spike Standard, Restek
Standards Environmental Analysis Standards
Five component standards are packaged in methanol in an ampule.
  • Contents of mixture: 4-Chloro-3-methylphenol, 2-Chlorophenol, 4-Nitrophenol, Pentachlorophenol, and Phenol
  • Highest concentrations commercially available
  • Reduces laboratory cost per sample extract
Each standard includes an ampule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty