Would you like to log in again?
Optic Prep™ Presaturated Wipes, ITW Chemtronics®
Supplier: ITW Chemtronics
Optic Prep™
CP410
47733-174CS 205.74 USD InStock
47733-174
Optic Prep™ Presaturated Wipes, ITW Chemtronics®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
High quality premoistened lens-grade tissue for cleaning optical surfaces.
  • Safely clean and polish optical surfaces, lenses, photographic equipment, CRT screens, and instruments
  • Ideal for field service kits
  • Nonabrasive
Quickly remove dust, oils, and contaminants. Solvent evaporates quickly and leaves no residue. Compatible with most plastics, glass, and metals. Each individual wipe is hermetically sealed to prevent evaporation and maintain solvent effectiveness.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty