Would you like to log in again?
Laparo® Laparoscopic Advanced Training Station
Supplier: AMERICAN 3B SCIENTIFIC SE
Laparo®
1021835
470355-812EA 15214.99 USD InStock
470355-812
Laparo® Laparoscopic Advanced Training Station
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Portable training station.
  • Mobile and stable construction
  • 12 precisely designed Trocars positions
  • Seven training modules included
  • Software includes video recording, time tracking and image adjustment
  • Multi language support
Independent training station with integrated touch-screen and foldable stand with height and worktop adjustment. Wheels with brakes allow to move the trainer fully, worktop has cut-outs to hold instruments.
Comes with a fully integrated touch-screen computer, box trainer, set of 6 modules and 4 laparoscopic instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty