Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Laparo® Apex VR Training Station
Supplier: AMERICAN 3B SCIENTIFIC SE
Laparo®
1023756
470355-766EA 206249.99 USD InStock
470355-766
Laparo® Apex VR Training Station
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Combines both physical training and VR.
  • Training on real laparoscopic instruments
  • Proficiency-based laparoscopic training
  • Precise fixation in a given position
  • Full HD (1920×1080) camera resolution zoom
  • Real instruments
Apex is a hybrid Laparoscopic trainer that combines virtual reality with physical skills training. Despite its complexity, the trainer offers extremely simple and intuitive manual skills and procedures training, a wide selection of training modules, scenarios for conducting training in virtual reality and a number of tutorials that guide the user step by step as one would do with a personal trainer.
While physical training is necessary to develop the workshop of manual skills, virtual reality helps in learning surgical procedures such as coagulation, clipping etc. while reflecting the conditions that arise during these procedures.
LAPARO Apex has been equipped with many specialized sensors that reflect the measured values, each parameter enables both the trainers and students to assess the effectiveness of the training and evaluate the learning progress.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty