Would you like to log in again?
CELLMASTER™ Cell Culture Roller Bottles, Polystyrene, Greiner Bio-One
Supplier: Greiner bio-one
CELLMASTER™
682012 682072 680048 682015 680058 682060 680068 680045
82051-010CS 355.85 USD InStock
82051-010 82051-022 89129-504 82051-020 82051-008 82051-016 82051-028 82051-006
CELLMASTER™ Cell Culture Roller Bottles, Polystyrene, Greiner Bio-One
Bottles Roller Bottles
Smooth surface cell culture roller bottles come in long and short forms, and are made of high grade polystyrene for stability and optical clarity, with physical surface treatment for improved cell adhesion.
  • Smooth surface
  • High-grade polystyrene construction
  • HDPE screw cap construction with either standard seal or vented filter
  • Free of RNase, DNase, and human DNA
  • Non-cytotoxic (EN ISO 10993) and non-pyrogenic
Two-phase blow-molding procedure creates seamless construction so that liquids will not leak.
Screw caps are high-density polyethylene and come in standard seal (non-vented) or vented filter. Caps feature short threads and large knurls for improved grip and easy opening with a 2/3 turn. Standard seal cap sits flush with rim of bottle to prevent significant gas exchange. Vented filter cap guarantees tightly closed, contamination-free cultivation. Polyethylene terephthalate (PET) and polytetrafluorethane (PTFE) filter has hydrophobic capillary pore membrane, with defined pore size 0.2 µm. Filter provides optimal protection against contamination and a high gaseous exchange rate which enables mass cultivation of cells using biocarbonate buffer and CO₂.
Colors: clear polystyrene roller bottle with black graduation print and white writing area, blue cap, white filter. Sterilized by irradiation (SAL 10⁻³) and tissue culture treated.
Temperature range: –20 to 60°C (–4 to 140°F)
For single use only; lot number and best-before dates located on label for maximum traceability.
Certifications: USP Class VI certified.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty