Would you like to log in again?
KLEENGUARD® A70 Chemical Spray Protection Apron, Kimberly-Clark
Supplier: Kimberly-Clark
KleenGuard®
97790 97790
89030-872CS 567.08 USD InStock
CA89030-872 89030-872
KLEENGUARD® A70 Chemical Spray Protection Apron, Kimberly-Clark
Disposable Garments
The KLEENGUARD® A70 line of apparel is designed to provide protection against chemical spray in hazardous waste remediation, environmental cleanup, petrochemical/oil refining, acid/caustic handling, biohazard cleanup, and pharmaceutical manufacturing applications
  • Seamless design
  • Soft and durable fabric
  • Wick-away lining for increased comfort
  • Neck loop and waist level ties provide secure fitting
NFPA 99 compliant for antistatic material.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty