Would you like to log in again?
CPP™ Vinyl Apron, Ansell
Supplier: Ansell
CPP™
950085
80093-262CS 297.3 USD InStock
80093-262
CPP™ Vinyl Apron, Ansell
Disposable Garments
Designed for use in food handling and preparation, industrial maintenance, and chemical processing, this versatile CPP™ brand apron offers medium- to heavy-duty splash protection.
  • Versatile and durable
  • Superior flexibility
  • Excellent chemical resistance
Die-cut from virgin vinyl, apron sType 56-015 provides superior flexibility and resistance to chemicals, fats, oils, grease, punctures, and abrasions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty