Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chlorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning
Synonyms: (4-Chlorophenyl)metaboric acidDihydroxy(4-chlorophenyl)borane(4-Chlorophenyl)boronic acid4-Chlorobenzeneboronic acid

10188-25G
77497-388EA 62.5 USD InStock
77497-388
4-Chlorophenylboronic acid ≥97%
4-Chlorophenylboronic acid
Caution: for Research Use Only


Formula: C₆H₆BClO₂
MW: 156.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00039137
CAS Number: 1679-18-1
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty